missing translation for 'onlineSavingsMsg'
Learn More

RON, MST1R Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

129.00 EUR - 349.00 EUR

Specifications

Antigen RON, MST1R
Clone 2D8D1
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30237137
View Documents
Proteintech
67954-1-IG-150UL
150 μL
ONLINE EXCLUSIVE PRICE
410.00 EUR
349.00 EUR / 150µL
Save 61.00 EUR
14% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30233193
View Documents
Proteintech
67954-1-IG-20UL
20 μL
ONLINE EXCLUSIVE PRICE
145.00 EUR
129.00 EUR / 20µL
Save 16.00 EUR
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The RON, MST1R antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RON, MST1R. This antibody recognizes human antigen. The RON, MST1R antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

RON, MST1R
1000 μg/mL
Monoclonal
Liquid
RUO
4486
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
2D8D1
Western Blot, ELISA
Unconjugated
Mouse
Human
CD136; CDw136; PTK8; RON
IgG1
Protein G purification
Monoclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.