missing translation for 'onlineSavingsMsg'
Learn More

RON, MST1R Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 67954-1-IG-150UL

 View more versions of this product

Product Code. 30237137

  • ONLINE EXCLUSIVE PRICE
    410.00 EUR
    349.00 EUR / 150µL
    Save 61.00 EUR
    14% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The RON, MST1R antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RON, MST1R. This antibody recognizes human antigen. The RON, MST1R antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

RON, MST1R
Monoclonal
1000 μg/mL
CD136; CDw136; PTK8; RON
Recombinant Protein
150 μL
Primary
Human
150-185 kDa
Liquid
Western Blot, ELISA
2D8D1
Unconjugated
Mouse
Protein G purification
RUO
4486
Store at -20°C. Stable for one year after shipment.
Monoclonal Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis