missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rociletinib, MedChemExpress
Description
Rociletinib (CO-1686) is an orally delivered kinase inhibitor that specifically targets the mutant forms of EGFR including T790M, and the Ki values for EGFRL858R/T790M and EGFRWT are 21.5 nM and 303.3 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Rociletinib |
| CAS | 1374640-70-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H28F3N7O3 |
| Quantity | 100 mg |
| Synonym | CO-1686 AVL-301 CNX-419 |
| Solubility Information | DMSO : ≥ 43 mg/mL (77.40 mM) |
| SMILES | C=CC(NC1=CC=CC(NC2=NC(NC3=CC=C(N4CCN(C(C)=O)CC4)C=C3OC)=NC=C2C(F)(F)F)=C1)=O |
| Molecular Weight (g/mol) | 555.55 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?