Learn More
MP Biomedicals RNase Erase™ Refill Bottle
Product Code. 11441722 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (Metric):
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11441722 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11441722 Supplier MP Biomedicals Supplier No. 04821684
  • ONLINE EXCLUSIVE PRICE
    88.20 EUR
    84.00 EUR / 250mL
    Save 4.20 EUR
    4% Off
 Restricted Item
Estimated Shipment: 22-02-2027
Removes RNase contamination from glass, plastic, pipettes, and equipment.
  • Removes RNase contamination
  • Clear, colorless liquid
  • Formulated using three ingredients known to be active against RNase
  • Effectively removes high levels of RNase contamination that similar products cannot
Specifications
Product Type RNase Decontaminant
Volume (Metric) 250 mL
For Use With (Application) Removing RNase contamination
Packaging Type Refill Bottle