Learn More
Cytiva RNA Homopolymers
Product Code. 11575115 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11575115 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11575115 Supplier Cytiva Supplier No. 27411001
  • ONLINE EXCLUSIVE PRICE
    315.15 EUR
    266.42 EUR / 100mg
    Save 48.73 EUR
    15% Off
 Restricted Item
In Stock
Template primers
These products are dispensed by weight of dry material, which includes polymer, residual salt, and residual water; content of polymer may vary from lot to lot.
TRUSTED_SUSTAINABILITY
Specifications
Product Type RNA Homopolymer
Content And Storage -20°C
Primer Poly(rA)
Quantity 100 mg
For Use With (Application) Evaluate the binding of potential therapeutic agents to mRNA in drug development research
Form Solid