Learn More
Ritiometan, TRC
Product Code. 30421647
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421647 1 g
30418169 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30421647 Supplier Toronto Research Chemicals Supplier No. TRCR5342501G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34914-39-1
Molecular Formula C7 H10 O6 S3
Molecular Weight (g/mol) 286.35
Synonym Acetic acid, 2,2',2' '-[methylidynetris(thio)]tris- (9CI), Acetic acid, S,S',S' '-methenyltris[mercapto- (1CI), Ritiometan
IUPAC Name 2-[bis(carboxymethylsulfanyl)methylsulfanyl]acetic acid
SMILES OC(=O)CSC(SCC(=O)O)SCC(=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C7H10O6S3/c8-4(9)1-14-7(15-2-5(10)11)16-3-6(12)13/h7H,1-3H2,(H,8,9)(H,10,11)(H,12,13)
Formula Weight 285.964
Chemical Name or Material Ritiometan