Learn More
Risperidone-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Risperidone-D4 |
| CAS | 1020719-76-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23 2H4 H23 F N4 O2 |
| Quantity | 2.5 mg |
| Synonym | Risperidone-D4, 3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl-1,1,2,2-d4]-6,7,8,9-tetrahydro-2-methyl-4H-Pyrido[1,2-a]pyrimidin-4-one |
| InChI Formula | InChI=1S/C23H27FN4O2/c1-15-18(23(29)28-10-3-2-4-21(28)25-15)9-13-27-11-7-16(8-12-27)22-19-6-5-17(24)14-20(19)30-26-22/h5-6,14,16H,2-4,7-13H2,1H3/i9D2,13D2 |
| SMILES | [2H]C([2H])(N1CCC(CC1)c2noc3cc(F)ccc23)C([2H])([2H])C4=C(C)N=C5CCCCN5C4=O |
| IUPAC Name | 2-methyl-3-[1,1,2,2-tetradeuterio-2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one |
| Molecular Weight (g/mol) | 414.51 |
| Show More |