Learn More
Risedronic Acid, TRC
Product Code. 30137948
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137948 250 mg 250mg
30136052 2.5 g 2.5g
30008828 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137948 Supplier Toronto Research Chemicals Supplier No. TRCR521505250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 105462-24-6
Molecular Formula C7 H11 N O7 P2
Molecular Weight (g/mol) 283.11
Synonym Phosphonic acid, P,P'-[1-hydroxy-2-(3-pyridinyl)ethylidene]bis-, Phosphonic acid, [1-hydroxy-2-(3-pyridinyl)ethylidene]bis- (9CI), P,P'-[1-Hydroxy-2-(3-pyridinyl)ethylidene]bis[phosphonic acid], 1-Hydroxy-2-(3-pyridinyl)ethylidene bisphosphonic acid, 1-Hydroxy-2-(3-pyridyl)ethylidene-1,1-diphosphonic acid, 1-Hydroxy-2-pyrid-3-ylethylidene-1,1-bisphosphonic acid, BPH 2, NE 58019, Risedronate, Risedronic acid
IUPAC Name (1-hydroxy-1-phosphono-2-pyridin-3-ylethyl)phosphonic acid
SMILES OC(Cc1cccnc1)(P(=O)(O)O)P(=O)(O)O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C7H11NO7P2/c9-7(16(10,11)12,17(13,14)15)4-6-2-1-3-8-5-6/h1-3,5,9H,4H2,(H2,10,11,12)(H2,13,14,15)
Formula Weight 283.0011
Percent Purity >95%
Chemical Name or Material Risedronic Acid
Grade HPLC