Learn More
Ripretinib, MedChemExpress
Product Code. 16464618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464618 10 mg 10mg
16460917 50 mg 50mg
16435466 100 mg 100mg
16427337 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16464618 Supplier Medchem Express Supplier No. HY112306
MedChemExpress Ripretinib (DCC-2618) is an orally bioavailable, selective KIT and PDGFRA switch-control inhibitor.
Ripretinib (DCC-2618) targets and binds to both wild-type and mutant forms of KIT and PDGFRA specifically at their switch pocket binding sites, thereby preventing the switch from inactive to active conformations of these kinases and inactivating their wild-type and mutant forms. Ripretinib (DCC-2618) also inhibits multiple other kinase targets, such as FLT3 and KDR (or VEGFR-2). DCC-2618 exerts antineoplastic effect and induces apoptosis.
Specifications
Chemical Name or Material Ripretinib
CAS 1442472-39-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H21BrFN5O2
Quantity 10 mg
Synonym DCC-2618
Solubility Information DMSO : 25 mg/mL (48.99 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C2=CC3=C(C=C(NC)N=C3)N(CC)C2=O)=C(Br)C=C1F)NC4=CC=CC=C4
Molecular Weight (g/mol) 510.36
Formula Weight 510.36
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease