Learn More
Epredia™ Neg-50™ Frozen Section Medium
Product Code. 12647756 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4 oz. per bottle
4 oz. per bottle, 2/cs.
4 oz. per bottle, 6/cs.
4 oz.per bottle
Color:
Blue
Clear
Five Colors plus Clear
Green
Orange
Pink
Yellow
Unit Size:
Pack of 2
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Color Unit Size
12647756 4 oz. per bottle, 6/cs. Clear Pack of 6
11912365 4 oz. per bottle, 2/cs. Clear Pack of 2
12678086 4 oz.per bottle Blue Pack of 2
12688086 4 oz. per bottle Green Pack of 2
12698086 4 oz. per bottle Orange Pack of 2
12608096 4 oz. per bottle Pink Pack of 2
12618096 4 oz. per bottle Yellow Pack of 2
12628096 4 oz. per bottle Five Colors plus Clear Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 12647756 Supplier Epredia™ Supplier No. 6506
  • ONLINE EXCLUSIVE PRICE
    141.00 EUR
    109.20 EUR / Pack of 6
    Save 31.80 EUR
    22% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Yield high-quality sections down to -50°C without chatter using water-soluble Epredia™ Neg-50™ Frozen Section Medium. Comes in clear and five additional colors with all leaving no residue after staining.
Yield high-quality sections down to -50°C without chatter using Epredia; Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
  • Buffered to ensure proper alkalinity (pH = 8.0); leaves no residue after staining
  • Specially formulated to prevent tissue separation from the slide
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Packaging Type 6/Cs.
Quantity 4 oz. per bottle, 6/cs.