missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ribonuclease A Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human Ribonuclease A.
Specifications
Specifications
| Assay Range | 62.5 pg/mL |
| Assay Sensitivity | 62.5-4000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes an antibody specific for Human Ribonuclease A. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1 |
| Gene ID (Entrez) | 6035 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction