missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin-5-phosphate, Sodium salt, MP Biomedicals™

Product Code. 11406202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406202 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11406202 Supplier MP Biomedicals Supplier No. 0210281605
  • ONLINE EXCLUSIVE PRICE
    61.75 EUR
    58.80 EUR / 5g
    Save 2.95 EUR
    4% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

USP grade photoinitiator for polymerization in acid or alkaline buffer systems.

Specifications

CAS 130-40-5
Color Orange
Molecular Formula C17H21N4NaO9P
MDL Number MFCD00065362,MFCD00150992
Quantity 5 g
InChI Key WXEASYCOEACRIK-UHFFFAOYNA-N
SMILES [Na].CC1=C(C)C=C2N(CC(O)C(O)C(O)COP(O)(O)=O)C3=NC(=O)NC(=O)C3=NC2=C1
IUPAC Name [(5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}-2,3,4-trihydroxypentyl)oxy]phosphonic acid sodium
Molecular Weight (g/mol) 479.34
PubChem CID 131673928
Formula Weight 478.3
Grade Not Available
Physical Form Powder
Chemical Name or Material Riboflavin-5-phosphate, sodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.