missing translation for 'onlineSavingsMsg'
Learn More

RGS16 Monoclonal Antibody (OTI2C9), TrueMAB™, OriGene

Product Code. 17591362
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17591362 30 μL 30µL
17571362 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17591362 Supplier Origene Technologies Supplier No. TA504308S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RGS16 Monoclonal antibody specifically detects RGS16 in Canine, Human, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen RGS16
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DPP9
Gene Accession No. Q86TI2
Gene Alias A28-RGS14; A28-RGS14P; hRGS-r; regulator of G-protein signaling 16; regulator of G-protein signalling 16; Retinally abundant regulator of G-protein signaling; retinal-specific RGS; Rgs14; RGS16; RGSR; RGS-R
Host Species Mouse
Immunogen Full length human recombinant protein of human DPP9 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301130, 485033, 91039
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.