Learn More
3M™ 6000 Series Reusable Full Face Mask Respirators
Product Code. 12352319 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12352319 Small Each
12362319 Medium Each
12372319 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12352319 Supplier 3M™ Supplier No. 7100015974
  • ONLINE EXCLUSIVE PRICE
    287.00 EUR
    149.33 EUR / Pack of 1
    Save 137.67 EUR
    47% Off
Estimated Shipment: 22-12-2026
Offers lightweight comfort and ease of use. 3M™ 6000 Series Reusable Full Face Mask Respirators have the 3M Bayonet Connection System, enabling connection to a broad range of twin lightweight filters to protect against gases, vapours and particulates.
  • Large lens for wide field of view and excellent visibility
  • Well-balanced design with silicone face seal for greater comfort and durability
  • Unique center adapter directs exhaled breath and moisture downward, reducing valve debris and facilitating quick and easy cleaning
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 136:1998 (Class 1)
Connection Type Bayonet
Color Light Gray
Material Silicone, Thermoplastic Elastomer
Product Type Reusable Full Face Mask Respirator
Size Small
Facepiece Type Full
For Use With (Application) For construction applications
For Use With (Equipment) 200 series filters and 600 series cartridges
Harness Style 4-point
Includes Bayonet connection, double gasket, phonic membrane, four-point head strap
Material (Lens) Polycarbonate Visor
Quantity 1 Ea.
Weight (Metric) 400 g