Learn More
RETSCH AS Series Sieve Shaker Accessory, Standard Clamping Device
Product Code. 10420081 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
AS Series
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10420081 AS Series Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10420081 Supplier RETSCH Supplier No. 32.662.0002
  • ONLINE EXCLUSIVE PRICE
    434.00 EUR
    423.53 EUR / Pack of 1
    Save 10.47 EUR
    2% Off
Estimated Shipment: 30-12-2026
Provide reliable, quick and convenient clamping of sieves in dry operation
Provide reliable, quick and convenient clamping of sieves in dry operation.
  • Threaded rods and clamping nuts
  • View cover
Specifications
Product Type Clamp
For Use With (Application) Reliable, quick, convenient clamping of sieves in dry operation; For Sieves: 4-8mm
For Use With (Equipment) AS Series