Learn More
RETSCH Aluminum Cups for Pellet Press PP 25
Product Code. 11964752 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric):
32 mm
40 mm
Includes:
1000 pieces
600 pieces
Unit Size:
Each
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Includes Unit Size
11964752 40 mm 600 pieces Each
11718224 32 mm 1000 pieces Pack of 1000
11728224 40 mm 600 pieces Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11964752 Supplier RETSCH Supplier No. 22.005.0002
  • ONLINE EXCLUSIVE PRICE
    682.00 EUR
    518.12 EUR / Pack of 1
    Save 163.88 EUR
    24% Off
Estimated Shipment: 29-01-2027
Specifications
Product Type Aluminum Cup
Diameter (Metric) 40 mm
For Use With (Equipment) Pellet Press PP 25
Includes 600 pieces
Material Aluminum