missing translation for 'onlineSavingsMsg'
Learn More

Retinol, MP Biomedicals™

Product Code. 11491761 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
25mg
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491761 100 mg 100mg
11481761 25 mg 25mg
11401771 250 mg 250mg
11411771 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11491761 Supplier MP Biomedicals Supplier No. 0219026880
  • ONLINE EXCLUSIVE PRICE
    60.65 EUR
    57.75 EUR / 100mg
    Save 2.90 EUR
    4% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 68-26-8
Molecular Formula C20H30O
Molecular Weight (g/mol) 286.459
MDL Number MFCD00001552
InChI Key FPIPGXGPPPQFEQ-OVSJKPMPSA-N
Synonym retinol, vitamin a, all-trans-retinol, vitamin a1, alphalin, axerophthol, vitamin a alcohol, oleovitamin a, chocola a, alphasterol
PubChem CID 445354
ChEBI CHEBI:17336
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraen-1-ol
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCO)C)C

Specifications

Color Orange
Quantity 100 mg
Formula Weight 286.5
Physical Form Powder
Chemical Name or Material Retinol, Molecular biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.