missing translation for 'onlineSavingsMsg'
Learn More

Retinol Acetate, Water Soluble, MP Biomedicals™

Product Code. 11411242 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411242 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411242 Supplier MP Biomedicals Supplier No. 0219931105
  • ONLINE EXCLUSIVE PRICE
    87.10 EUR
    82.95 EUR / 5mg
    Save 4.15 EUR
    4% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Often used in cell culture applications. Retinol Acetate, Water Soluble, MP Biomedicals (aka Vitamin A) is found in classical media formulations, such as M-199; NCTC-109 and William’s Medium E. Typical working concentration is ca. 3.44mg of retinol acetat

Vitamin A is a component of serum used to supplement classical media. Consequently, most serum-containing cell culture systems receive some level of vitamin A. The oxidized form of retinol is retinoic acid, which has hormone-like growth factor and differentiation activity for epithelial and related cell types, especially neural cells. Retinoic acid induces embryonic stem cells to differentiate towards neural cells.

Specifications

CAS 127-47-9
Melting Point 57°C to 58°C (literature)
Molecular Formula C22H32O2
Quantity 5 mg
Synonym retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650
Solubility Information Soluble in chloroform (50mg/mL, clear yellow solution); absolute ethanol: soluble 25mg/mL (Lit.)
InChI Key QGNJRVVDBSJHIZ-QHLGVNSISA-N
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
Molecular Weight (g/mol) 328.496
PubChem CID 638034
ChEBI CHEBI:32095
Formula Weight 328.5g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.