missing translation for 'onlineSavingsMsg'
Learn More

Retinoic acid, MedChemExpress

Product Code. 16421746
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421746 500 mg 500mg
16471429 1 g 1g
16426516 100 mg 100mg
16428727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16421746 Supplier Medchem Express Supplier No. HY14649

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Retinoic acid is a metabolite of vitamin A that plays important roles in cell growth, differentiation, and organogenesis. Retinoic acid is a natural agonist of RAR nuclear receptors, with IC50s of 14 nM for RARα/β/γ. Retinoic acid bind to PPARβ/δ with Kd of 17 nM. Retinoic acid acts as an inhibitor of transcription factor Nrf2 through activation of retinoic acid receptor alpha.

Retinoic acid is a metabolite of vitamin A that plays important roles in cell growth, differentiation, and organogenesis. Retinoic acid is a natural agonist of RAR nuclear receptors, with IC50s of 14 nM for RARα/β/γ. Retinoic acid bind to PPARβ/δ with Kd of 17 nM. Retinoic acid acts as an inhibitor of transcription factor Nrf2 through activation of retinoic acid receptor alpha.

Specifications

Chemical Name or Material Retinoic acid
CAS 302-79-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H28O2
Quantity 500 mg
Synonym Vitamin A acid all-trans-Retinoic acid ATRA
Solubility Information DMSO : 50 mg/mL (166.42 mM; ultrasonic and warming and heat to 60°C)
SMILES CC1(C)C(/C=C/C(C)=C/C=C/C(C)=C/C(O)=O)=C(C)CCC1
Molecular Weight (g/mol) 300.44
Formula Weight 300.44
Percent Purity 96.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.