missing translation for 'onlineSavingsMsg'
Learn More

Retinoic Acid, All Trans Isomer, MP Biomedicals™

Product Code. 11441771 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441771 500 mg 500mg
11431771 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441771 Supplier MP Biomedicals Supplier No. 02190269.5
  • ONLINE EXCLUSIVE PRICE
    196.00 EUR
    185.85 EUR / 500mg
    Save 10.15 EUR
    5% Off
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mediates the functions of vitamin A required for growth and development. Retinoic Acid, All Trans Isomer, MP Biomedicals has been used in various differentiation protocols, including B-cells, T cells and neurons, and is applied clinically to treat cancer

Retinoic acid (RA) is a vitamin A-derived, non-peptidic, small lipophilic molecule. It affects gene transcription and modulates a wide variety of biological processes, including cell proliferation and differentiation, including apoptosis.

  • Retinoic Acid mediated gene transcription depends on the rate of transport of Retinoic Acid to target cells and the timing of exposure of Retinoic Acid to RARs (Retinoic Acid Receptors) in the target tissues.
  • All-trans-Retinoic acid (ATRA) is a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR).
  • The bound RAR and RXR act as transcription factors that regulate the growth and differentiation of both normal and malignant cells.
  • Cytochromes P450 (CYPs) catalyze the 4-hydroxylation of ATRA.
  • Retinoic acid primes embryonic stem cells to become neurons.

Chemical Identifiers

CAS 302-79-4
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.44
MDL Number MFCD00001551
InChI Key SHGAZHPCJJPHSC-YCNIQYBTSA-N
Synonym retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra
PubChem CID 444795
ChEBI CHEBI:15367
SMILES C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O

Specifications

Melting Point 173°C to 183°C
Quantity 500 mg
Solubility Information Soluble in methanol, chloroform (40mg/mL) or acetone (30mg/mL), slightly soluble in ethanol.
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Formula Weight 300.4
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.