Learn More
Retaspimycin Hydrochloride, MedChemExpress
Product Code. 16486447
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486447 5 mg 5mg
16480097 50 mg 50mg
16454786 100 mg 100mg
16430979 1 mg 1mg
16413718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16486447 Supplier Medchem Express Supplier No. HY10210
MedChemExpress Retaspimycin Hydrochloride is a potent inhibitor of Hsp90 with EC50s of 119 nM for both Hsp90 and Grp9.
Retaspimycin Hydrochloride is a potent inhibitor of Hsp90 with EC50s of 119 nM for both Hsp90 and Grp9.
Specifications
Chemical Name or Material Retaspimycin Hydrochloride
CAS 857402-63-2
Color Pink
Purity Grade Notes Research
Molecular Formula C31H46ClN3O8
Quantity 5 mg
Synonym IPI-504
Solubility Information 1M HCl : 100 mg/mL (160.21 mM; ultrasonic and adjust pH to 1 with 1M HCl) ∣DMSO : 60 mg/mL (96.13 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(NC1=C2O)/C(C)=C/C=C\[C@H](OC)[C@H](/C(C)=C/[C@@H]([C@H]([C@H](C[C@@H](CC2=C(C(O)=C1)NCC=C)C)OC)O)C)OC(N)=O.Cl
Molecular Weight (g/mol) 624.17
Formula Weight 624.17
Percent Purity 98.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease