missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein G Antibody (11-2-D2), DyLight 594, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Respiratory Syncytial Virus Glycoprotein G Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein G in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Functional Assay
Specifications
Specifications
| Antigen | Respiratory Syncytial Virus Glycoprotein G |
| Applications | Western Blot, ELISA, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Clone | 11-2-D2 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Gradient-purified RSF-44 virus (subgroup A) UV inactivated for 20 minutes at 20C |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?