Learn More
3M™ Respirator Carry Cases
Código de producto. 15222800 Tienda 3M Productos
Cambiar vista
Click to view available options
Includes:
Adjustable shoulder strap, name card viewing window
Tamaño de la unidad:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Includes Tamaño de la unidad
15222800 Adjustable shoulder strap, name card viewing window Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15222800 Proveedor 3M™ N.º de proveedor 7000087291
  • ONLINE EXCLUSIVE PRICE
    89.00 EUR
    52.79 EUR / Pack of 1
    Save 36.21 EUR
    40% Off
Estimated Shipment: 25-11-2026
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
A large and robust case to carry full face masks and spare filters.
  • Fits any full face mask
  • Robust and durable design
  • Adjustable shoulder strap
  • Name card window included
  • Provides convenient and clean storage between use
TRUSTED_SUSTAINABILITY
Specifica
Color Blue
Includes Adjustable shoulder strap, name card viewing window
For Use With (Application) Storage and transport of full face mask respirators
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.