Learn More
Resorcinol, Sigma-Aldrich™
Product Code. 15816750
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15816750 250 g 250g
15801781 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15816750 Supplier Sigma Aldrich Supplier No. 8223030250
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-46-3
Specifications
Melting Point 109°C to 111°C
Density 1.28 g/cm3 at 20°C
Boiling Point 281°C/1013 hPa
Molecular Formula C6H4(OH)2
MDL Number MFCD00002269
Quantity 250 g
Synonym Resorcinol, 1,3-Dihydroxybenzene, Resorcinol
Molecular Weight (g/mol) 110.11
Grade Research