Learn More
Resorcinol Bis(diphenyl phosphate), TRC
Product Code. 30011085
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011085 100 mg 100mg
30006262 1 g 1g
30003906 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011085 Supplier Toronto Research Chemicals Supplier No. TRCR145300100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57583-54-7
Molecular Formula C30 H24 O8 P2
Molecular Weight (g/mol) 574.45
Synonym Phosphoric acid, P,P'-1,3-phenylene P,P,P',P'-tetraphenyl ester, Phosphoric acid, 1,3-phenylene tetraphenyl ester (9CI), 1,3-Phenylene bis(diphenyl phosphate), ADK Stab PFP, ADK Stab PFR, BPHPPO, CR 733S, Fyrolflex RDP, LDP 301, Mark PFK, Nonnen R 0111-10, PFR, PhireGuard RDP, Reofos RDP, Resorcinol bis(diphenyl phosphate), Resorcinol tetraphenyl diphosphate, Tetraphenyl m-phenylene bisphosphate, Tetraphenyl m-phenylene diphosphate, Tetraphenyl resorcinol diphosphate, WSFR-RDP, m-Phenylene bis(diphenyl phosphate)
IUPAC Name (3-diphenoxyphosphoryloxyphenyl) diphenyl phosphate
SMILES O=P(Oc1ccccc1)(Oc2ccccc2)Oc3cccc(OP(=O)(Oc4ccccc4)Oc5ccccc5)c3
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C30H24O8P2/c31-39(33-25-14-5-1-6-15-25,34-26-16-7-2-8-17-26)37-29-22-13-23-30(24-29)38-40(32,35-27-18-9-3-10-19-27)36-28-20-11-4-12-21-28/h1-24H
Formula Weight 574.0946
Chemical Name or Material Resorcinol Bis(diphenyl phosphate)