Learn More
Electrothermal™ Replacement Spare Heater Element
Product Code. 11768432
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11768432 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11768432 Supplier Electrothermal™ Supplier No. 1008647
  • ONLINE EXCLUSIVE PRICE
    502.00 EUR
    374.65 EUR / Pack of 1
    Save 127.35 EUR
    25% Off
Estimated Shipment: 19-11-2026
Specifications
Capacity (Metric) 1000 mL
For Use With EMEA3/6/1000
Voltage 230 V
Wattage 300w
Product Type Replacement Spare Heater Element
Frequency 50 Hz