Learn More
Merck Replacement Parts for Stirred Cells
Description
- Small Replacement Parts for 8000 Series Stirred Cells include 2 O-rings (Mfr. Nos. 2122S, 2118S, 2015S), tubing (Mfr. No. 4500202), tubing fitting assembly (Mfr. No. 15025AM, and membrane holder (Mfr. No. 2214801)
- Large Replacement Parts for Stirred Cells include one each: cap assembly, stirrer assembly and base
Specifications
Specifications
| Includes | Cap assembly (1524901), stirrer assembly (15247) and base (210000) |
| Type | Replacement Part |
| For Use With (Equipment) | Amicon 8003 Series Stirred Cell |