Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
Product Code. 15145283 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
ClickSeal Biocontainment Lid
O-ring
For Use With (Equipment):
MicroClick 24 x 2 Rotor
MicroClick 24 x 2 Rotor Lid
MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) Unit Size
15145283 O-ring MicroClick 24 x 2 Rotor Lid Each
15165273 ClickSeal Biocontainment Lid MicroClick 24 x 2 Rotor Each
15125263 O-ring MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors Each
15114933 ClickSeal Biocontainment Lid MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15145283 Supplier Thermo Scientific™ Supplier No. 75003405
  • ONLINE EXCLUSIVE PRICE
    145.00 EUR
    133.00 EUR / Pack of 1
    Save 12.00 EUR
    8% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
A range of options for secure closure on microcentrifuge rotors to help contain samples.
  • O-rings for inner and outer seals
  • ClickSeal lids for optimal biocontainment
  • Available for a variety of sizes and models
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MicroClick 24 x 2 Rotor Lid
Product Type O-ring