Learn More
Klever Replacement Head (Narrow Cut Head)
Product Code. 17779007
Change view
Click to view available options
Quantity:
25/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17779007 25/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17779007 Supplier Klever Supplier No. KCJXH20X
  • ONLINE EXCLUSIVE PRICE
    171.47 EUR
    138.23 EUR / Pack of 50
    Save 33.24 EUR
    19% Off
Estimated Shipment: 12-01-2027
Replacement Head (narrow cut head) for use with Klever X-Change Safety Cutters, is best for all-around cutting applications.
  • Recessed blade reduces cut injuries and damaged goods.
  • Cuts boxes, film, tape, and thousands more applications.
  • Ergonomic soft touch handle reduces hand fatigue and stress.
  • Innovative metal tape splitter
  • Highest safety level
  • Also available in replaceable narrow and multipurpose wide heads
  • Our antimicrobial Kleen™ safety cutters utilize an antimicrobial material that inhibits the growth of bacteria that may affect this product.
  • The antimicrobial properties do not protect users or others against bacteria, viruses, germs, or other disease organisms.
  • Clean and wash this product thoroughly per your industry's guidelines.
  • The active antimicrobial component is part of the formulation, so it won't wash off or wear away. This is not a coating or treatment; it's an integral ingredient.
Specifications
Blade Material Carbon Steel
Product Type Replacement Head
For Use With (Application) Cutting
For Use With (Equipment) Kleen™ XChange KCJ-XC-20X
Disposable Single-use
Sterility Non-sterile
Quantity 25/Pk.