Learn More
Electrothermal™ Replacement Digi-Mantle Heater Cartridge
Beschreibung
- For use with the OMCA Digital Controller.
- Stir and heat round bottom flasks up to an element temperature of 450°C
- Uses precise electronic control to give an accurate measure of temperature and stirring.
- Modular design allows heating cartridges of different sizes to be interchanged using the same OMCA digital controller as a base
- Cost-effective and versatile
Spezifikation
Spezifikation
| Capacity (Metric) | 250 mL |
| For Use With | OMCA Digi-Mantle |
| Product Type | Replacement Heating Cartridge |