Learn More
Grant Instruments™ Replacement Base Tray for Unstirred Water Baths
Product Code. 11446417
Change view
Click to view available options
For Use With (Equipment):
Sub Aqua Pro 12L
Sub Aqua Pro 18/26L
Sub Aqua Pro 2L
Sub Aqua Pro2S, 5L JB Nova/Academy
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11446417 Sub Aqua Pro 18/26L Each
11436417 Sub Aqua Pro 12L Each
11486407 Sub Aqua Pro 2L Each
11496407 Sub Aqua Pro2S, 5L JB Nova/Academy Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11446417 Supplier Grant Instruments™ Supplier No. AQBT26
  • ONLINE EXCLUSIVE PRICE
    68.00 EUR
    60.43 EUR / Pack of 1
    Save 7.57 EUR
    11% Off
Estimated Shipment: 21-12-2026
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments Replacement Base Tray for Unstirred Water Baths promote thermal convection in the bath.
  • For use with Sub Aqua Pro Series Unstirred Water Bath
  • Choose a rack that fits 2, 5, 12, or 26L tank size
  • Racks are supplied as standard with purchase of a Sub Aqua Pro Unstirred water Bath
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Sub Aqua Pro 18/26L
Type Base Tray
Material Polycarbonate