Learn More
Rephile™ RephiDuo S Pack 2 Kit
Product Code. 15469749 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15469749 Water Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15469749
  • ONLINE EXCLUSIVE PRICE
    459.02 EUR
    372.37 EUR / Pack of 1
    Save 86.65 EUR
    18% Off
 Restricted Item
Estimated Shipment: 04-05-2027
Add to basket
Add to basket
This product is designed to remove the ionic and organic contaminants below trace level to match the water quality required for your application.
This product is designed to remove the ionic and organic contaminants below trace level to match the water quality required for your application. A 0.2 μm capsule filter is included to remove bacteria and particles above 0.2 μm size. For use with DI feed water.
RephiDuo S Pack 2 Kit for older MilliQ Simplicity™ and Synergy™ systems. Comparable to SIMPAKKD2.
Compatible with following Millipore™ systems:
  • Milli-Q Simplicity™
  • Milli-Q Synergy™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit