Learn More
Rephile™ RephiDuo L Pack L1
Product Code. 15469729 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15469729 Water Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15469729
  • ONLINE EXCLUSIVE PRICE
    412.16 EUR
    334.21 EUR / Pack of 1
    Save 77.95 EUR
    18% Off
 Restricted Item
Estimated Shipment: 17-05-2027
Add to basket
Add to basket
This product is for additional/upstream pretreatment (extra protection) and designed to remove most of the particles, organic compounds and chlorine from tap water to protect the TL1 Pretreatment Pack and the RO membranes.
RephiDuo L Pack L1 for large Elix™ systems. Comparable to PRPK0L0S1.
Compatible with following Millipore™ systems:
  • Elix Clinical 35/70™
  • Elix 20/35/70/100™
  • RiOs 30/50/100/150/200™
  • Elix Gulfstream Clinical™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit