Learn More
Gilac™ Removable Pulling Bar for 4, 5, or 6 Wheel Dollies
Product Code. 17938761
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
17938761 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17938761 Supplier Gilac™ Supplier No. G170428
  • ONLINE EXCLUSIVE PRICE
    159.60 EUR
    115.06 EUR / Pack of 1
    Save 44.54 EUR
    27% Off
Estimated Shipment: 30-11-2026
Removable Pulling Bar for 4, 5, or 6 Wheel Dollies makes moving heavy loads easier. It's suitable for GILAC 4, 5, and 6 wheel dollies.
  • Removable pulling bar for 600 x 400 (4 and 6 wheels) dollies and round (5 wheels) dolly.
  • Makes moving heavy loads easier.
  • Suitable for GILAC 4, 5, and 6 wheel dollies.
  • Enables effortless dolly handling, both pulling and pushing.
  • Systematically returns to the upright position.
  • The mount remains on the dolly, the arm of the pulling bar is removable
  • New detachable retaining pin fastening system.
Specifications
Holds Dolly
Material Stainless Steel
For Use With (Equipment) GILAC 4, 5, and 6 Wheel Dollies