missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Remodelin

Description
Remodelin is a N-acetyltransferase 10 (NAT10) inhibitor. Improves nuclear shape in HGPS (Hutchinson-Gilford progeria syndrome) cells and lamin A/C depleted cells.
Specifications
Specifications
| Chemical Name or Material | 4-[2-(2-Cyclopentylidenehydrazinyl)-4-thiazolyl]benzonitrile |
| CAS | 949912-58-7 |
| Quantity | 50 mg |
| Target | Other Transferase Inhibitors |
| Molecular Formula | C15H14N4S |
| Formula Weight | 282.36 |
| Purity | >99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?