missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Remodelin
GREENER_CHOICE

Product Code. 18762459
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18762459 10 mg 10mg
18777048 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18762459 Supplier Tocris Bioscience™ Supplier No. 5193/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-acetyltransferase 10 (NAT10) inhibitor

Remodelin is a N-acetyltransferase 10 (NAT10) inhibitor. Improves nuclear shape in HGPS (Hutchinson-Gilford progeria syndrome) cells and lamin A/C depleted cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[2-(2-Cyclopentylidenehydrazinyl)-4-thiazolyl]benzonitrile
CAS 949912-58-7
Quantity 10 mg
Target Other Transferase Inhibitors
Molecular Formula C15H14N4S
Formula Weight 282.36
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.