Learn More
Thermo Scientific™ Remel™ BactiCard™ Neisseria
Product Code. 13255659
Change view
Click to view available options
Quantity:
25 Tests/Kit
Unit Size:
25 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13255659 25 Tests/Kit 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13255659 Supplier Thermo Scientific™ Supplier No. R21110
  • ONLINE EXCLUSIVE PRICE
    222.00 EUR
    177.45 EUR / 25 tests
    Save 44.55 EUR
    20% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Für den Erwerb von Giften gemäß §35 Chemikaliengesetz 1996 (ChemG 1996) durch Betriebe und sonstige berufsmäßige Verwenderinnen/Verwender wird eine Giftbezugsbescheinigung benötigt.
Uses enzyme technology for presumptive identification of Neisseria spp. and Moraxella catarrhalis on selective media.
Thermo Scientific™ Remel BactiCard Neisseria uses enzyme technology for presumptive identification of pathogenic Neisseria spp. and Moraxella catarrhalis isolated on selective media.
TRUSTED_SUSTAINABILITY
Spezifikation
Description BactiCard™ Neisseria
CE Marker Yes
Detectable Analytes Neisseria species , M. catarrhalis
Type Bacticard Neisseria
For Use With (Application) Enzyme Immunoassay Detection
Quantity 25 Tests/Kit
Target Organism Class Pathogenic Neisseria Spp., Moraxella Catarrhalis