Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent
Product Code. 11993322
Change view
Click to view available options
Description:
Polyvalent (group A - G)
Polyvalent (group A - S)
Unit Size:
2mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
11993322 Polyvalent (group A - S) 2mL
12627499 Polyvalent (group A - G) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11993322 Supplier Thermo Scientific™ Supplier No. R30858201
  • ONLINE EXCLUSIVE PRICE
    155.00 EUR
    142.33 EUR / 2mL
    Save 12.67 EUR
    8% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Remel Agglutinating Sera, Salmonella O is used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Control Sets Positive
Description Polyvalent (group A - S)
Detectable Analytes Salmonella O Antigen
Type Agglutinating Sera
Material Polyvalent
Organism Group Group A-S
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella O