missing translation for 'onlineSavingsMsg'
Learn More

Virkon™ Rely+On™ Disinfectant (Nordics)

Product Code. 31903153
Change view
Click to view available options
Packaging:
1 Tub
12 Tubs/Box
20 Tubs/Box
Form:
Granular Powder
Tablet
Unit Size:
Pack of 12
Pack of 20
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Form unitSize
31903153 20 Tubs/Box Tablet Pack of 20
31903150 12 Tubs/Box Tablet Pack of 12
31903151 1 Tub Granular Powder Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Read instructions before use or contact manufacturer for the latest Health and Safety advice
Product Code. 31903153 Supplier Virkon™ Supplier No. 350009

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Read instructions before use or contact manufacturer for the latest Health and Safety advice

Clean and disinfect medical, laboratory, public and work surfaces and equipment in a single, one-step operation. Virucidal Disinfectant Powder provides the widest proven spectrum of any disinfectant.

  • Use biocides safely. Always read the label and product information before use
  • Proven effective in independent tests against all 18 virus families affecting humans and animals, including AIDS (HIV) and Hepatitis B
  • Excellent bactericide and fungicide
  • Can be applied in powder form direct to spillages of blood and other body fluids
  • Easily dissolves to make a liquid disinfectant
  • Odourless, non-irritant to skin and eyes, non-toxic vapour phase, exceptionally low oral toxicity
  • Under COSHH requirements, no maximum exposure limit is required
  • Non-corrosive, non-bleaching and biodegradable
  • Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva can be controlled by correct application

Specifications

Product Type Disinfectant Tablet
Color-coded Yes
Form Tablet
Product Line Cleanware
Scent Lemon
Sterility Sterile
Appearance Red-Brown
Autoclavable Autoclavable
Color Red
Disposable Single-use
Filtered Filtered
Flammable No
Packaging 20 Tubs/Box
Quantity 10 x 5 g
Show More Show Less
compliance-icons
Product Identifier
  • RELYON VIRKON NORDIC
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P310-Immediately call a POISON CENTER/doctor/.

UK biocidal regulations (HSE / BPR)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.