missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relaxin R1/LGR7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
230.00 EUR - 372.00 EUR
Specifications
| Antigen | Relaxin R1/LGR7 |
|---|---|
| Dilution | Western Blot reported in scientific literature (PMID 31218471), Immunohistochemistry, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 31218471) |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18130682
|
Novus Biologicals
NBP2-23674SS |
0.025 mL |
230.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18197751
|
Novus Biologicals
NBP2-23674 |
0.1 mL |
372.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Relaxin R1/LGR7 Polyclonal specifically detects Relaxin R1/LGR7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifica
| Relaxin R1/LGR7 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| Crude antibody whole antisera with 50% Glycerol with 0.025% Sodium Azide | |
| leucine-rich repeat-containing G protein-coupled receptor 7, Leucine-rich repeat-containing G-protein coupled receptor 7, LGR7.1, LGR7.10, LGR7LGR7.2, MGC138347, MGC142177, Relaxin family peptide receptor 1, relaxin receptor 1, relaxin/insulin-like family peptide receptor 1, RXFPR1 | |
| RXFP1 | |
| IgG | |
| Unpurified | |
| The amino acid sequence used as immunogen for this RXFP1 antibody is 100% homologous in Human, Chimp and Orangutan, and 83% homologous in Horse, Bovine, Dog and Cat. |
| Western Blot reported in scientific literature (PMID 31218471), Immunohistochemistry, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 31218471) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| NP_067647.2 | |
| 59350 | |
| Within the range of amino acids 694-732 of human RXFP1 protein were used as the immunogen for this antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto