missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relaxin R1/LGR7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-23674
This item is not returnable.
View return policy
Description
Relaxin R1/LGR7 Polyclonal specifically detects Relaxin R1/LGR7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Relaxin R1/LGR7 | |
| Polyclonal | |
| Western Blot reported in scientific literature (PMID 31218471), Immunohistochemistry, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 31218471) | |
| NP_067647.2 | |
| RXFP1 | |
| Within the range of amino acids 694-732 of human RXFP1 protein were used as the immunogen for this antibody. | |
| 0.1 mL | |
| GPCR | |
| 59350 | |
| Human | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Crude antibody whole antisera with 50% Glycerol with 0.025% Sodium Azide | |
| leucine-rich repeat-containing G protein-coupled receptor 7, Leucine-rich repeat-containing G-protein coupled receptor 7, LGR7.1, LGR7.10, LGR7LGR7.2, MGC138347, MGC142177, Relaxin family peptide receptor 1, relaxin receptor 1, relaxin/insulin-like family peptide receptor 1, RXFPR1 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this RXFP1 antibody is 100% homologous in Human, Chimp and Orangutan, and 83% homologous in Horse, Bovine, Dog and Cat. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction