Learn More
BD Biosciences™ Regulatory T-Cell Function Kit (RUO/GMP)
Description
- Naturally occurring human CD4+ regulatory T cells (Tregs) are critical mediators of self-tolerance and suppress aberrant or inappropriate immune responses that can lead to conditions such as type 1 diabetes, rheumatoid arthritis, and graft-vs-host disease.
- The BD FastImmune™ kit measures expression of the activation markers CD154 and/or CD69 on effector T cells.
- Reduced expression of these markers in the presence of Tregs can indicate Treg suppressive capacity.
- The assay is performed in 96-well microplates using peripheral blood mononuclear cells (PBMCs) activated in the presence and absence of Tregs.
- It is a short-term assay that can be set up and analyzed in approximately one day.
- The T-cell markers, CD3 and CD4, are included in the kit for T-cell gating purposes, and CD25 is included for defining the Treg population.
- Contents: Regulatory T-Cell function cocktail (CD3 PerCP-Cy™5.5, CD4 FITC, CD25 PE); CD69 PE-Cy™7; CD154 APC; EDTA.
Specifications
Specifications
| For Use With (Equipment) | Flow cytometer |
| Formulation | CD154, APC, CD4 FITC/CD25 PE/CD3 PerCP-CY 5.5, CD69, PE-CY7, EDTA |
| Includes | CD154, APC, CD4 FITC/CD25 PE/CD3 PerCP-CY 5.5, CD69, PE-CY7, EDTA |
| Quantity | 50 Tests |
| Regulatory Status | RUO/GMP |
| Content And Storage | Store reagents at 2°C to 8°C. Conjugated forms should not be frozen and should be protected from exposure to light. |