Learn More
Grant Instruments™ Refrigerated Immersion Coolers
Product Code. 11476487
Change view
Click to view available options
Temperature Range:
-15°C to +40°C
0°C to 40°C
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range Unit Size
11476487 -15°C to +40°C Each
11466487 0°C to 40°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11476487 Supplier Grant Instruments™ Supplier No. C2GR
  • ONLINE EXCLUSIVE PRICE
    3145.00 EUR
    2929.41 EUR / Pack of 1
    Save 215.59 EUR
    6% Off
 Restricted Item
Estimated Shipment: 05-03-2027
Refrigerated immersion coolers consists of a cooling coil connected to a refrigeration unit by a flexible pipe. Extract heat continuously, with the bath control unit controlling temperature.
  • C1GR 0ºC to 40ºC
  • The cooling coil can be continuously immersed in liquids up to 100ºC with the cooler switched off and may be used to cool liquid down from 100ºC but it is not designed for continuous operation above 40ºC.
  • C2GR: -15ºC to 40ºC
TRUSTED_SUSTAINABILITY
Specifications
Description 460 x 305 x 225 mm (D x W x H)
Cooling Capacity 400w at 20°C, 320w at 0°C, 170w at -10°C
Temperature Range -15°C to +40°C
Length (Metric) Hose 925 mm
Electrical Requirements 120/230 V
Voltage 120/230 V