Learn More
Tork Reflex Centrefeed Paper Towel and Dispenser M4
Product Code. 30243834 Shop All Tork Products
Change view
Click to view available options
Model:
Manual
Mobile
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
30243834 Manual Each
30243767 Mobile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30243834 Supplier Tork Supplier No. 473186
  • ONLINE EXCLUSIVE PRICE
    70.98 EUR
    65.44 EUR / Pack of 1
    Save 5.54 EUR
    7% Off
Estimated Shipment: 21-12-2026
The portable Tork Reflex Single Sheet Centrefeed system is the ideal hand and surface wiping solution for professional environments on the move.
  • Reduce the risk of cross-contamination - our single-sheet dispensing system means you touch only what you take.
  • Meet the challenges of tough environments with this robust centrefeed dispenser.
  • Boost hygiene levels - the dispenser fully encloses the roll to protect paper from dirt.
  • Keep track of consumption and never be left without paper with the run out indicator.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hard Roll Towel Dispenser
Color White, Blue
Alternate Part Number 7322540953916
Holds Hard Roll Towel
Material Plastic
Mounting Freestanding
No. of Compartments 1
Product Style Top Loading
Model Manual