Learn More
Labcon™ Refillable Box for 192 Pipette Stacking Racks
Product Code. 17557876 Shop All Labcon Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17557876 White Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17557876 Supplier Labcon™ Supplier No. 12151200009
  • ONLINE EXCLUSIVE PRICE
    12.60 EUR
    12.28 EUR / Pack of 5
    Save 0.32 EUR
    2% Off
Estimated Shipment: 02-12-2026
Refillable box for storing racked pipette tips.
  • Refillable rack with hinged clear lid
  • Accepts two Labcon™ 8 x 24 pipette floating racks
  • Freezable to -80°C
  • Made in USA
Specifications
Certifications/Compliance ISO 9001:2015, FDA CFR21 GMP
Autoclavable Autoclavable
Color White
Material Polypropylene
Shape Rectanglular
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) Floating Racks
No. per Case 50/Cs.
No. per Pack 5/Pk.
Quantity 5 Boxes/Pk., 10 Pk./Cs.
Includes Hinged Box without Racks
Product Type Freezer Storage Box
Array 8 x 24