Learn More
Redox Standard Solution, +358mV, Chem-Lab
Product Code. 11923093
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11923093 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11923093 Supplier Chem-Lab Supplier No. CL03.1804.0500
  • ONLINE EXCLUSIVE PRICE
    52.90 EUR
    41.30 EUR / 500mL
    Save 11.60 EUR
    21% Off
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Ensure that electrodes are calibrated correctly for maximum accuracy. Redox Standard Solution, +358mV, Chem-Lab is used with Redox/ORP electrodes with a silver/silver chloride wire in saturated potassium chloride. Millivolt output: 358mV at 25°C. Store away from light.
Redox (reduction–oxidation) reactions cause an electron transfer process that can be measured using a Redox or ORP (oxidation-reduction potential) electrode. The milllivolt (mV) output directly correlates with the redox potential of the reaction. Redox standard solutions help calibrate and verify the measurements of Redox electrodes.
Millivolt Output Specifications:
  • 10°C—376 mV
  • 12°C—373 mV
  • 14°C—371 mV
  • 16°C—368 mV
  • 18°C—366 mV
  • 20°C—364 mV
  • 22°C—361 mV
  • 24°C—359 mV
  • 25°C—358 mV
  • 26°C—357 mV
  • 28°C—355 mV
  • 30°C—352 mV
  • 32°C—350 mV
  • 34°C—347 mV
  • 36°C—345 mV
  • 38°C—343 mV
  • 40°C—340 mV
Specifications
Density 1.02gm/mL
Conductivity 358mV at 25°C
For Use With (Equipment) Redox and ORP Electrodes
Quantity 500 mL
Concentration or Composition (by Analyte or Components) K3(Fe(CN)6)
K4(Fe(CN)6) • 3H2O
H2O