Learn More
Redox Standard Solution, 220mV, Crison Instruments™
Product Code. 12071452
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12071452 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12071452 Supplier Crison Instruments Supplier No. LZW9400.99
 Restricted Item
Estimated Shipment: 21-10-2026