Learn More
Tocris Bioscience™ RECTAS
Description
RECTAS (rectifier of aberrant splicing) is a pre-mRNA splicing modulator of the IKBKAP (inhibitor of κ light polypeptide gene enhancer in B-cells, kinase complex associated protein) gene. It corrects aberrant IKBKAP splicing in vitro in cellular models of familial dysautonomia and in vivo in transgenic mice. In combination with Phenylbutyric acid (Cat. No. 2682), RECTAS rescues PARK7 aberrant splicing and neuronal cell loss in a midbrain organoid model of familial Parkinson's disease. Orally bioavailable and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-N-(2-furanylmethyl)-7 H-purin-6-amine |
| CAS | 101862-47-9 |
| Quantity | 10 mg |
| Target | DNA, RNA and Protein Synthesis Compounds |
| Molecular Formula | C10H8ClN5O |
| Purity | 0.98 |