Learn More
R&D Systems™ Reconstitution Buffer 2 (BSA/PBS)
Product Code. 30002147
Click to view available options
Quantity:
2 mL
Unit Size:
Each
Product Code. 30002147
Brand: R&D Systems™ RB02
0.1% Bovine Serum Albumin in PBS
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity. Refer to the specific antibody or protein product data sheet for the recommended reconstitution concentrations.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 mL