Learn More
Recombinant VHH Alpaca, anti-Mouse, Unconjugated, Clone: CTK0106, Proteintech ChromoTek
Product Code. 17343493
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17343493 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17343493 Supplier Chromotek Supplier No. smsG2bCys22500
 Restricted Item
In Stock
Add to basket
Add to basket
Alpaca Monoclonal Antibody
500 μg Nano-Secondary™ for mouse IgG2b: alpaca monoclonal Nanobody, Fc-specific VHH, for 2x Cys conjugation, clone [CTK0106].
Specifications
Antigen Recombinant VHH
Applications Immunofluorescence, Western Blot, Flow Cytometry
Classification Monoclonal
Clone CTK0106
Conjugate Unconjugated
Host Species Alpaca
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse